


1,2-Diethoxyethane 99%
:629-14-1
:
:C₆H₁₄O₂
:MFCD00009253
:118.18 g/mol
:121.4 °C (1013 hPa)
:–74 °C
:0.8417 g/cm³
:19 °C
:1153
:3,II
:GHS07
:GHS08
:GHS02
:Ambient
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3910 to 1.3930 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥98.0 %
Frequently Bought Together