ContactUsLinkComponent
Chloro(1,5-cyclooctadiene)rhodium(I) dimer (>40.8% Rh)
Irritant
Chloro(1,5-cyclooctadiene)rhodium(I) dimer (>40.8% Rh)
Certificates
undefined
Irritant
Chloro(1,5-cyclooctadiene)rhodium(I) dimer (>40.8% Rh)
Specification Test Results
  • Appearance:
    Orange to orange-brown Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Rhodium (Rh):
    ≥40.8 %