
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64536-78-3
:
Formula:C₃₁H₅₀F₆IrNP₂
MDL Number:MFCD00075097
Melting Pt:175 °C (dec.)
:804.90 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Orange Powder
- Infrared spectrum:Conforms
- Carbon (C):45.6 to 47.0 %
Specification Test Results
Frequently Bought Together