
cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate 99%
Catalog # ACRO363300010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Black Powder or crystals
- Infrared spectrum:Conforms
- Elemental Analysis:≥98.5 % (ex Ru)
Specification Test Results
About this item
:
:520.376 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:15746-57-3
Formula:C₂₀H₂₀Cl₂N₄O₂Ru
MDL Number:MFCD00015504