Order Entry
Czech Republic
ContactUsLinkComponent
(2R,4S)-N-Boc-4-hydroxypiperidine-2-carboxylic acidmethyl ester 95%, 98% ee
(2R,4S)-N-Boc-4-hydroxypiperidine-2-carboxylic acidmethyl ester 95%, 98% ee
Catalog # ACRO362970010
CAS Number:  
undefined
(2R,4S)-N-Boc-4-hydroxypiperidine-2-carboxylic acidmethyl ester 95%, 98% ee
Catalog # ACRO362970010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to amber Viscous liquid or waxy mass
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥94 %
  • Enantiomeric excess:
    ≥97.5 % (GC)

Specification Test Results

About this item

 :
 :259.302 g/mol
Storage Temperature:Refrigerator
CAS Number:321744-26-7
Formula:C₁₂H₂₁NO₅
MDL Number:MFCD02259744