Order Entry
Czech Republic
ContactUsLinkComponent
Methyllithium 1.6 M in diethyl ether (± 5% w/v), AcroSeal®
Irritant
Corrosive
Flammable
Methyllithium 1.6 M in diethyl ether (± 5% w/v), AcroSeal®
Certificates
undefined
Irritant
Corrosive
Flammable
Methyllithium 1.6 M in diethyl ether (± 5% w/v), AcroSeal®

Specification Test Results

  • Appearance:
    Colorless to yellow Solution (May develop some turbidity or precipitate)
  • Titration Total base:
    4.5 to 5.5 %
  • HPLC:
    ≥97.5 %

Specification Test Results

Frequently Bought Together