




Ethylaluminum sesquichloride 0.4 M in hexane, AcroSeal®
:12075-68-2
:
:C₆H₁₅Al₂Cl₃
:MFCD00044852
:247.499 g/mol
:0.701 g/cm³
:–22 °C
:Ambient
:3399
:4.3,I
:GHS07
:GHS08
:GHS05
:GHS09
:GHS02
- Appearance:Clear colorless Solution
- Assay:14.5 to 15.5 % (on Al basis)
- Solubility:(1% in water) Clear colorless to light yellow
- HPLC:≥80 % (tylosin A)
Frequently Bought Together