
(1S,4R)-N-Boc-1-aminocyclopent-2-ene-4-carboxylic acid 95%, 98% ee
Catalog # ACRO361820010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥94 %
- Enantiomeric excess:≥97.5 % (HPLC)
Specification Test Results
About this item
CAS Number:151907-80-1
:
Formula:C₁₁H₁₇NO₄
MDL Number:MFCD00211288
:227.26 g/mol
Melting Pt:152 °C
:GHS07
Storage Temperature:Refrigerator