

1,1-Di-(tert-butylperoxy)-3,3,5-trimethylcyclohexane 75% in aromatic free mineral spirit
Katalogové číslo ACRO349772500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 mL
- Seq:2590
Specifications
Specification Test Results
- Appearance:Clear solution
- Titration Iodometric:74 to 76 %
- Color scale:≤50 APHA
- Impurity:≤0.8 % Hydroperoxides (as TBHP)
Specification Test Results
About this item
:
:302.455 g/mol
Boiling Pt:–20 °C
Melting Pt:–20 °C
Density:0.87 g/cm³
Flash Pt:62 °C
UN:3103
ADR:5.2,
:GHS02
:GHS08
Storage Temperature:Refrigerator
CAS Number:6731-36-8
Formula:C₁₇H₃₄O₄
MDL Number:MFCD00084801