
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:138124-32-0
:
Formula:C₃₆H₅₂ClMnN₂O₂
MDL Number:MFCD02101664
:635.21 g/mol
Melting Pt:330…332 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark brown Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Elemental Analysis:≥96.0 %
Specification Test Results
Frequently Bought Together