Order Entry
Czech Republic
ContactUsLinkComponent
 
9-Benzhydrylidene-10-anthrone 95%
 
undefined
9-Benzhydrylidene-10-anthrone 95%

 

  • Appearance:
    Yellow to beige Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    202 to 212 °C
  • HPLC:
    ≥94 %

 

 

Frequently Bought Together