
Ethyl-2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate 99%
Catalog # ACRO251280100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:114 to 117 °C
- GC:≥98.5 %
Specification Test Results
About this item
:
:225.31 g/mol
Melting Pt:113…117 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
CAS Number:4506-71-2
Formula:C₁₁H₁₅NO₂S
MDL Number:MFCD00005862