

Cyclopropanecarboxaldehyde 98%
:1489-69-6
:
:C₄H₆O
:MFCD00012261
:70.09 g/mol
:98…101 °C
:0.938 g/cm³
:7 °C
:Ambient
:1989
:3,II
:GHS07
:GHS02
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4285 to 1.4305 (20°C, 589 nm)
- GC:≥97.5 %
Frequently Bought Together