


Hydrochloric acid 4 N in 1,4-dioxane/water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Melting Pt:-30 °C
Density:1.1 g/cm³
Flash Pt:12 °C
UN:2924
ADR:3,II
:36.46 g/mol
:GHS08
:GHS05
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light yellow Solution
- Strength:3.95 to 4.40 N
Specification Test Results
Frequently Bought Together