1-(Mesitylene-2-sulphonyl)-3-nitro-1,2,4-triazole 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:74257-00-4
:
Formula:C₁₁H₁₂N₄O₄S
MDL Number:MFCD00009754
:296.301 g/mol
Melting Pt:132…134 °C
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to yellow Powder or crystals
- Infrared spectrum:Conforms
- Melting point:131 to 139 °C
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together