Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
Calcium sulphate, Drierite™ 8 mesh for drying, with indicator
 
undefined
 
 
 
Calcium sulphate, Drierite™ 8 mesh for drying, with indicator

 

  • Appearance:
    Blue Irregular granules
  • Water:
    ≤0.3 %
  • Particle size:
    8 mesh
  • Composition:
    ≥98 % (CaSO4) Typical

 

 

Frequently Bought Together