Order Entry
Czech Republic
ContactUsLinkComponent
2-Dimethylaminoethyl methacrylate 99% stabilised
Irritant
2-Dimethylaminoethyl methacrylate 99% stabilised
Certificates
undefined
Irritant
2-Dimethylaminoethyl methacrylate 99% stabilised

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4390 to 1.4410 (20°C, 589 nm)
  • Water:
    ≤0.1 % (K.F.)
  • GC:
    ≥98.5 %
  • Stabilizer:
    750 to 2000 ppm MEHQ

Specification Test Results

Related Information

Frequently Bought Together