Sodium-DL-3-hydroxybutyrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:150-83-4
:
Formula:C₄H₈O₃
MDL Number:MFCD00016716
:126.09 g/mol
Melting Pt:161…167 °C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:161 to 167 °C (Dec)
- Titration with HClO4:≥98.5 %
Specification Test Results
Frequently Bought Together