Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:50525-27-4
:
Formula:C₃₀H₃₆Cl₂N₆O₆Ru
MDL Number:MFCD00149670
:748.62 g/mol
Melting Pt:>300 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:Orange to red Crystalline powder or crystals
- Elemental Analysis:≥97.5 %
Specification Test Results
Frequently Bought Together