Order Entry
Czech Republic
ContactUsLinkComponent
L-Phenylalanine ethyl ester hydrochloride 99%
Irritant
L-Phenylalanine ethyl ester hydrochloride 99%
Certificates
undefined
Irritant
L-Phenylalanine ethyl ester hydrochloride 99%

Specification Test Results

  • Appearance:
    White Crystalline powder or needles
  • Infrared spectrum:
    Conforms
  • Melting point:
    154 to 157 °C
  • Specific optical rotation:
    +32.2° to +35.2° (20 °C, 589 nm) (c=2, C2H5OH)
  • Titration with HClO4:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together