Hydroxy naphthol blue disodium salt ACS
:165660-27-5
:
:C₂₀H₁₂N₂O₁₁S₃·2Na
:MFCD00004075
:598.479 g/mol
:274 °C
:GHS07
:Ambient
- Appearance:Blue to dark purple Crystalline powder or crystals
- Infrared spectrum:Conforms
- Suitability:(for Ca determination) Passes test
Frequently Bought Together