ContactUsLinkComponent
DL-2,3-Diaminopropionic acid hydrochloride 99%
undefined
DL-2,3-Diaminopropionic acid hydrochloride 99%
  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥98.5 %
Frequently Bought Together