

Sodium borohydride 0.5 M in diglyme, AcroSeal®
Catalog # 191131000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless Solution
- Strength:0.45 to 0.55 M
- Solution contains:18.915 g NaBH4/l C6H14O3
Specification Test Results
About this item
CAS Number:16940-66-2
:
Formula:NaBH₄
MDL Number:MFCD00003518
:37.83 g/mol
Density:0.945 g/cm³
Flash Pt:69 °C
ADR:4.3,I
:GHS07
:GHS08
Storage Temperature:Ambient