Ethyl cellulose, 22 cps 48% (ethoxyl content)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9004-57-3
:
MDL Number:MFCD00131037
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Crystalline powder or granules
- Infrared spectrum:Conforms
- Assay:48 to 49.5 % (Ethoxyl)
- Viscosity:18 to 24 cP (5% soln. in 80/20 toluene/ethanol, 25°C)
Specification Test Results
Frequently Bought Together