Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
Mercurochrome Hg 24-27% 24-27% Hg
 
undefined
 
 
 
Mercurochrome Hg 24-27% 24-27% Hg

 

  • Appearance:
    Green Glistening crystals or granules or scales
  • Assay:
    24 to 27 % (Hg)

 

 

Frequently Bought Together