Poly(vinyl chloride), carboxylated, M.W. ∼220000 aver. carboxyl content 1.8 wt%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25702-80-1
:
Formula:(C₅H₇ClO₂)n
MDL Number:MFCD00084460
Density:1.39 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Water:5.0 to 11.0 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together