Order Entry
Czech Republic
ContactUsLinkComponent
5-Bromothiophene-2-carbaldehyde 97%
Irritant
5-Bromothiophene-2-carbaldehyde 97%
Certificates
undefined
Irritant
5-Bromothiophene-2-carbaldehyde 97%

Specification Test Results

  • Appearance:
    Clear yellow to brown Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6380 to 1.6410 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together