

2-(4-Aminophenyl)ethylamine 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13472-00-9
:
Formula:C₈H₁₂N₂
MDL Number:MFCD00008194
:136.20 g/mol
Boiling Pt:103 °C (.3 mmHg)
Melting Pt:28 °C
Density:1.057 g/cm³
Flash Pt:>112 °C
Storage Temperature:Ambient
UN:3259
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear colorlesss to yellow-brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5890 to 1.5930 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
Frequently Bought Together