

ɑ-Chloralose 98+% (max. 20% beta-anomer)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15879-93-3
:
Formula:C₈H₁₁Cl₃O₆
MDL Number:MFCD00005542
:309.52 g/mol
Melting Pt:176…182 °C
UN:2811
:GHS06
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Melting point:178 to 183 °C
- GC:≥98.0 % (alpha + beta)
- Specific optical rotation:+15° to +19° (20 °C, 589 nm) (C=2, Ethanol)
Specification Test Results
Frequently Bought Together