

4,5,6,7-Tetrahydroisobenzofuran-1,3-dione 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2426-02-0
:
Formula:C₈H₈O₃
MDL Number:MFCD00005917
:152.15 g/mol
Melting Pt:70…74 °C
UN:2698
ADR:8,III
:GHS08
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Melting point:70 to 74 °C
- Titration with NaOH:≥97.5 %
Specification Test Results
Frequently Bought Together