
2-Hydroxypyrimidine hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:38353-09-2
:
Formula:C₄H₄N₂O
MDL Number:MFCD00012781
:132.55 g/mol
Melting Pt:203…205 °C (dec.)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light yellow to beige Crystalline solid
- Infrared spectrum:Conforms
- Titration with HClO4:≥97.5 %
- NMR (1H-NMR):Consistent with structure
- Titration with NaOH:≥98.0 %
Specification Test Results
Frequently Bought Together