Order Entry
Czech Republic
ContactUsLinkComponent
Nε-carbobenzoxy-L-lysine 98%
Nε-carbobenzoxy-L-lysine 98%
Catalog # ACRO108360250
CAS Number:  
undefined
Nε-carbobenzoxy-L-lysine 98%
Catalog # ACRO108360250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to almost white Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤1 % (K.F.)
  • Sulfated ash:
    ≤0.2 %
  • HPLC:
    ≥97.5 %
  • Specific optical rotation:
    +12.0° to +16.0° (20°C, 589 nm) (c=1, 1N HCl)
  • Titration with HClO4:
    ≥97.5 %
  • Sulfate (SO4):
    ≤300 ppm
  • Ammonium (NH4):
    ≤200 ppm
  • Iron (Fe):
    ≤10 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Other amino acids:
    ≤0.5 % (TLC)

Specification Test Results

About this item

 :
 :280.32 g/mol
Melting Pt:259 °C (dec.)
Storage Temperature:Ambient
CAS Number:1155-64-2
Formula:C₁₄H₂₀N₂O₄
MDL Number:MFCD00002638