2,2,4-Trimethyl-1,3-pentanediol diisobutyrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6846-50-0
:
Formula:C₁₆H₃₀O₄
MDL Number:MFCD00059267
:286.41 g/mol
Boiling Pt:280 °C (760 mmHg)
Melting Pt:–70 °C
Density:0.94 g/cm³
Flash Pt:128 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4330 to 1.4350 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together