Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5574-97-0
:
Formula:C₁₆H₃₈NO₄P
MDL Number:MFCD00064526
:339.46 g/mol
Melting Pt:148…151 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- UV:(0.1 % in water) 1 cm cell
- Titration with NaOH:≥98.5 %
Specification Test Results
Frequently Bought Together