

13-cis-Retinoic acid 99%
Catalog # ACRO418975000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mg
- Seq:2725
Specifications
Specification Test Results
- Appearance:Yellow to orange Crystalline powder
- Infrared spectrum:Conforms
- Melting point:174 to 177 °C
- HPLC:≥98.5 %
- Specific optical rotation:+28° to +31° (20 °C, 589 nm) (c=1, H2O)
- Titration with HClO4:97.5 to 102.5 % (On anhydrous basis)
Specification Test Results
About this item
:
:300.44 g/mol
Melting Pt:174…177 °C
:GHS07
:GHS08
Storage Temperature:Freezer
CAS Number:4759-48-2
Formula:C₂₀H₂₈O₂
MDL Number:MFCD00079542