(R)-1-Phenyl-1,3-propanediol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:103548-16-9
:
Formula:C₉H₁₂O₂
MDL Number:MFCD00145213
:152.193 g/mol
Melting Pt:62…66 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline solid
- Infrared spectrum:Conforms
- Melting point:62 to 66 °C
- GC:≥97.5 %
- Specific optical rotation:+53° to +56° (22 °C, 589 nm) (c=1, 1,2-dichlorethane)
Specification Test Results
Frequently Bought Together