Order Entry
Czech Republic
ContactUsLinkComponent
Methyl orange-xylene cyanol solution in water, pure
Methyl orange-xylene cyanol solution in water, pure
Catalog # ACRO414675000
CAS Number:  
undefined
Methyl orange-xylene cyanol solution in water, pure
Catalog # ACRO414675000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Dark blue-green Solution
  • Visual transition interval:
    from pH 3.2 (purple) to pH 4.2 (green)
  • GC:
    ≥88 % may occur as tautomers

Specification Test Results

About this item

 :
 :881.942 g/mol
Density:1 g/cm³
Storage Temperature:Ambient
CAS Number:62758-15-0
Formula:C₃₉H₄₁N₅Na₂O₁₀S₃
MDL Number:MFCD00077269