DL-Histidine monohydrochloride monohydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123333-71-1
:6459-59-2
Formula:C₆H₉N₃O₂·HCl·H₂O
MDL Number:MFCD00151028
:209.63 g/mol (191.62 anhy)
Melting Pt:253 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Water:7 to 10 %
- Sulfated ash:≤0.2 %
- Solubility:(3 % in H2O) Clear colorless to
- Titration with HClO4:97.5 to 102.5 %
- Sulfate (SO4):≤300 ppm
- Arsenic (As):≤1 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
- Impurity:≤0.02 % Ammonium salts (NH4)
- Other amino acids:(TLC) None
Specification Test Results
Frequently Bought Together