ContactUsLinkComponent
N,N-Dimethyl-1,4-phenylenediamineoxalate 99%
Toxic
N,N-Dimethyl-1,4-phenylenediamineoxalate 99%
Certificates
undefined
Toxic
N,N-Dimethyl-1,4-phenylenediamineoxalate 99%
Specification Test Results
  • Appearance:
    Beige to grey to brown Powder and/or chunks
  • Infrared spectrum:
    Conforms
  • Titration with NaOH:
    ≥98.5 %
  • HPLC:
    ≥97.0 %