


N,N-Dimethyl-n-octadecylamine 89% tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:124-28-7
:
Formula:C₂₀H₄₃N
MDL Number:MFCD00048496
:297.57 g/mol
Boiling Pt:347 °C (760 mmHg)
Melting Pt:15…20 °C
Density:0.8 g/cm³
Flash Pt:155 °C
UN:2735
ADR:8,II
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid after melting
- Infrared spectrum:Conforms
- Water:≤0.5 % (K.F.)
- GC:≥84 % (C18)
- Titration:≥95 % (Tertiary amine (MW 297))
Specification Test Results
Frequently Bought Together