Order Entry
Czech Republic
ContactUsLinkComponent
Pentasodium diethylenetriaminepentaacetate 40% in aqueous solution tech.
Corrosive
Irritant
Health
Pentasodium diethylenetriaminepentaacetate 40% in aqueous solution tech.
Certificates
undefined
Corrosive
Irritant
Health
Pentasodium diethylenetriaminepentaacetate 40% in aqueous solution tech.

Specification Test Results

  • Appearance:
    Clear solution
  • Color scale:
    ≤150 APHA
  • pH:
    11.0 to 12.0 (1 % in water)
  • Titration:
    39 to 41 %

Specification Test Results

Related Information

Frequently Bought Together