Order Entry
Czech Republic
ContactUsLinkComponent
 
Dextran for biochemistry, high fraction
 
undefined
Dextran for biochemistry, high fraction

 

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤9.0 %
  • Sulfated ash:
    ≤2.0 %
  • UV:
    (6 % in water)
  • Viscosity:
    0.4 to 0.5 (1 % at 37°C)
  • Average molecular weight:
    250000
  • pH:
    4.5 to 7.0 (6 % in water)

 

 

Frequently Bought Together