Order Entry
Czech Republic
ContactUsLinkComponent
trans-1,4-Cyclohexanedimethanol 98%
Irritant
trans-1,4-Cyclohexanedimethanol 98%
Catalog # ACRO406062500
CAS Number:  
undefined
Irritant
trans-1,4-Cyclohexanedimethanol 98%
Catalog # ACRO406062500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    61 to 66 °C
  • GC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :144.21 g/mol
Melting Pt:61…66 °C
Density:1.02 g/cm³
Flash Pt:>109 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:3236-48-4
Formula:C₈H₁₆O₂
MDL Number:MFCD00066360