Order Entry
Czech Republic
ContactUsLinkComponent
Cellulose acetate butyrate (35 - 39% butyryl)
Cellulose acetate butyrate (35 - 39% butyryl)
Catalog # ACRO404255000
CAS Number:  
undefined
Cellulose acetate butyrate (35 - 39% butyryl)
Catalog # ACRO404255000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    195 to 205 °C
  • Viscosity:
    17 to 28 s
  • Average molecular weight:
    70000 (Mn)
  • Acetyl content:
    12 to 15 %
  • Hydroxyl content:
    1.8 % Typical
  • Butyryl content:
    35 to 39 %
  • Free acid (CH3COOH):
    ≤0.03 %

Specification Test Results

About this item

 :
 :1923.90 g/mol
Melting Pt:127…240 °C
Storage Temperature:Ambient
CAS Number:9004-36-8
Formula:C₈₄H₁₃₀O₄₉
MDL Number:MFCD00081513