3,3'-Carbonylbis(7-diethylaminocoumarin) 99%
Catalog # ACRO404050050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Yellow to orange Crystalline powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥98.5 %
- UV Lambda max:447 to 451 nm (in toluene)
- Molar absorption coefficient:≥80000 l/(mol.cm) (at lambda max.)
Specification Test Results
About this item
:
:460.53 g/mol
Melting Pt:214 °C
Storage Temperature:Ambient
CAS Number:63226-13-1
Formula:C₂₇H₂₈N₂O₅
MDL Number:MFCD00050047