
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14337-53-2
:
Formula:C₁₅H₁₇BrN₄O
MDL Number:MFCD00006255
:349.23 g/mol
Melting Pt:155…160 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Red to dark red to purple red Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Solubility:complete dissolution within max 5 min
- Titration with HClO4:≥97.5 %
Specification Test Results
Frequently Bought Together