Acid orange 5 indicator grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:554-73-4
:
Formula:C₁₈H₁₄N₃NaO₃S
MDL Number:MFCD00038130
:375.38 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Ochre to orange Powder
- Infrared spectrum:Conforms
- Visual transition interval:from pH 1.4 (red) to pH 2.6 (yellow)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together