4-Nitrophenyl-β-D-glucuronide 99+%
:10344-94-2
:
:C₁₂H₁₃NO₉
:MFCD00036210
:315.23 g/mol
:Refrigerator
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Water:≤8 % (K.F.)
- Solubility:(1 % in water) Clear colorless to light yellow
- HPLC:≥99.0 %
- Specific optical rotation:-107° to -113° (20°C, 589 nm) (c=1, H2O)
Frequently Bought Together