Order Entry
Czech Republic
ContactUsLinkComponent
(+)-Dibenzoyl-L-tartaric anhydride 98%
Corrosive
(+)-Dibenzoyl-L-tartaric anhydride 98%
Certificates
undefined
Corrosive
(+)-Dibenzoyl-L-tartaric anhydride 98%

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    192 to 196 °C
  • Titration by coupling:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together