Diethyl (4-nitrobenzyl)phosphonate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2609-49-6
:
Formula:C₁₁H₁₆NO₅P
MDL Number:MFCD00047747
:273.225 g/mol
Boiling Pt:151 °C (.1 mmHg)
Density:1.24 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear yellow to orange to dark red Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5220 to 1.5260 (20°C, 589 nm)
- NMR:≥97.5 %
Specification Test Results
Frequently Bought Together